missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-5-trimethylsilyl-4-pentyne, 97%, Thermo Scientific Chemicals

50.65 € - 174.87 €

Chemical Identifiers

CAS 77113-48-5
Molecular Formula C8H15ClSi
Molecular Weight (g/mol) 174.74
MDL Number MFCD00671353
InChI Key UPVHIBLXQNNITH-UHFFFAOYSA-N
Synonym 5-chloro-1-pentynyl trimethylsilane, 1-chloro-5-trimethylsilyl-4-pentyne, silane, 5-chloro-1-pentynyl trimethyl, acmc-1bffc, 5-chloro-1-trimethylsilylpent-1-yne, 5-chloropent-1-ynyl trimethyl silane, 5-chloropent-1-yn-1-yl trimethylsilane, silane, 5-chloro-1-pentyn-1-yl trimethyl
PubChem CID 2757942
SMILES C[Si](C)(C)C#CCCCCl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15440158
View Documents
Thermo Scientific Alfa Aesar
H53393.06
5 g
50.65 €
5g
Please to purchase this item. Need a web account? Register with us today!
15450158
View Documents
Thermo Scientific Alfa Aesar
H53393.14
25 g
174.87 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

77113-48-5
174.74
UPVHIBLXQNNITH-UHFFFAOYSA-N
2757942
C8H15ClSi
MFCD00671353
5-chloro-1-pentynyl trimethylsilane, 1-chloro-5-trimethylsilyl-4-pentyne, silane, 5-chloro-1-pentynyl trimethyl, acmc-1bffc, 5-chloro-1-trimethylsilylpent-1-yne, 5-chloropent-1-ynyl trimethyl silane, 5-chloropent-1-yn-1-yl trimethylsilane, silane, 5-chloro-1-pentyn-1-yl trimethyl
C[Si](C)(C)C#CCCCCl

Specifications

75°C to 77°C (21mmHg)
C8H15ClSi
MFCD00671353
5-chloro-1-pentynyl trimethylsilane, 1-chloro-5-trimethylsilyl-4-pentyne, silane, 5-chloro-1-pentynyl trimethyl, acmc-1bffc, 5-chloro-1-trimethylsilylpent-1-yne, 5-chloropent-1-ynyl trimethyl silane, 5-chloropent-1-yn-1-yl trimethylsilane, silane, 5-chloro-1-pentyn-1-yl trimethyl
C[Si](C)(C)C#CCCCCl
174.74
174.75
1.456
1-Chloro-5-trimethylsilyl-4-pentyne
77113-48-5
Cl(CH2)3C≡CSi(CH3)3
65°C (149°F)
UPVHIBLXQNNITH-UHFFFAOYSA-N
(5-chloropent-1-yn-1-yl)trimethylsilane
2757942
97%
0.977
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.