missing translation for 'onlineSavingsMsg'
Learn More

1-Chloromethyl-1H-1,2,4-triazole hydrochloride, 96%, Thermo Scientific Chemicals

Product Code. 15498655
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84387-61-1
Molecular Formula C3H4ClN3
Molecular Weight (g/mol) 117.54
MDL Number MFCD00964947
InChI Key PXNMFQMREJNQQT-UHFFFAOYSA-N
PubChem CID 3743882
SMILES ClCN1C=NC=N1

Specifications

Melting Point 138°C to 145°C
Quantity 1 g
Sensitivity Hygroscopic
Synonym 1-chloromethyl-1h-1,2,4-triazole, 1-chloromethyl-1,2,4-triazole, 1h-1,2,4-triazole,1-chloromethyl, 1h-1,2,4-triazole, 1-chloromethyl, 1-chloro-methyl-1h-1,2,4-triazole
IUPAC Name 1-(chloromethyl)-1H-1,2,4-triazole
Formula Weight 154
Percent Purity 96%
Chemical Name or Material 1-Chloromethyl-1H-1,2,4-triazole hydrochloride
compliance-icons
Product Identifier
  • 1-Chloromethyl-1H-1,2,4-triazole hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Chloromethyl-1H-1,2,4-triazole hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'