missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate, 95%

Product Code. 10657062 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10657062

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2491-17-0 | C21H33N3O4S | 423.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-17-0
Molecular Formula C21H33N3O4S
Molecular Weight (g/mol) 423.57
MDL Number MFCD00011979
InChI Key GBCAVSYHPPARHX-UHFFFAOYSA-M
Synonym cme-carbodiimide, morpho cdi, 1-cyclohexyl 3-2-morpholinoethyl carbodiimide methotosylate, 4-2-cyclohexylimino methylene amino ethyl-4-methylmorpholin-4-ium 4-methylbenzenesulfonate, cmc, 1-cyclohexyl-3-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, n-cyclohexyl-n'-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, cmc n-cyclohexyl-n-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, chm van, 4-5-cyclohexyl-3,5-diazapenta-3,4-dienyl-4-methylmorpholine, 4-methylbenzene sulfonic acid
PubChem CID 17220
IUPAC Name N'-cyclohexyl-N-[2-(4-methylmorpholin-4-ium-4-yl)ethyl]methanediimine;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 110.0°C to 116.0°C
Color White to Yellow
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Fieser 01,181; 11,151
Solubility Information Solubility in water: soluble in water
Formula Weight 423.57
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.