missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Deazaadenosine, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Inhibitor of adenosine deaminase (Ki = 0.66 μM). Demonstrates antitumor activity in a range of leukemia cell lines.
Specifications
Specifications
CAS | 14432-09-8 |
Molecular Formula | C11H14N4O4 |
Quantity | 10 mg |
Synonym | 1-deazaadenosine, 1-deaza-adenosine, 1da, 1add, 2r,3r,4s,5r-2-7-aminoimidazo 4,5-b pyridin-3-yl-5-hydroxymethyl oxolane-3,4-diol, 1-deaza-a, d03plv, adenosine-n,1,3-15n3, 3-, a-d-ribofuranosyl-3h-imidazo 4,5-b pyridin-7-amine |
InChI Key | NVUDDRWKCUAERS-PNHWDRBUSA-N |
SMILES | C1=CN=C2C(=C1N)N=CN2C3C(C(C(O3)CO)O)O |
IUPAC Name | (2R,3R,4S,5R)-2-(7-aminoimidazo[4,5-b]pyridin-3-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
Molecular Weight (g/mol) | 266.257 |
PubChem CID | 159738 |
Formula Weight | 266.25 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'