missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Decyne, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.
Solubility
Not miscible in water.
Notes
Incompatible with strong oxidizing agents. Store at 0-6°C.
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.
Solubility
Not miscible in water.
Notes
Incompatible with strong oxidizing agents. Store at 0-6°C.

Specifications
Specifications
Melting Point | -44°C |
Density | 0.767 |
Boiling Point | 174°C to 176°C |
Flash Point | 50°C (122°F) |
Odor | Petrol-like |
Refractive Index | 1.427 |
Quantity | 25 g |
UN Number | UN3295 |
Beilstein | 1236372 |
Solubility Information | Not miscible in water. |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction