missing translation for 'onlineSavingsMsg'
Learn More

1-Docosanol, 98%, Thermo Scientific Chemicals

Product Code. 10272262
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661-19-8
Molecular Formula C22H46O
Molecular Weight (g/mol) 326.61
MDL Number MFCD00002939
InChI Key NOPFSRXAKWQILS-UHFFFAOYSA-N
Synonym 1-docosanol, behenyl alcohol, docosanol, behenic alcohol, docosyl alcohol, n-docosanol, abreva, tadenan, lidavol, lanette 22
PubChem CID 12620
ChEBI CHEBI:31000
IUPAC Name docosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCO

Specifications

CAS Min % 97.5
CAS Max % 100
Melting Point 68.0°C to 72.0°C
Color White
Flash Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3(CH2)21OH
Quantity 25 g
Beilstein 01, 431
Merck Index 15, 3444
Solubility Information Solubility in water: insoluble.
Viscosity 7.5 mPa.s (80°C)
Formula Weight 326.61
Percent Purity 98%
Physical Form Pellets or Tablets
Chemical Name or Material 1-Docosanol, 0.98
Show More Show Less
Product Identifier
  • 1-Docosanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.