missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Dodecanol, ACS Reagent, ≥98%, Honeywell Riedel-de Haën™
ACS Reagent,≥98.0%
108.53 € - 266.30 €
Chemical Identifiers
CAS | 112-53-8 |
---|---|
Molecular Formula | C12H26O |
Molecular Weight (g/mol) | 186.34 |
MDL Number | MFCD00004753 |
InChI Key | LQZZUXJYWNFBMV-UHFFFAOYSA-N |
Synonym | 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol |
PubChem CID | 8193 |
ChEBI | CHEBI:28878 |
IUPAC Name | dodecan-1-ol |
SMILES | CCCCCCCCCCCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15695550
|
Honeywell-Riedel-de Haen
443816-500G |
500 g | Poly Bottle | N/A | |||||
15685550
|
Honeywell-Riedel-de Haen
443816-1KG |
1 kg | Poly Bottle |
266.30 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
112-53-8 | |
186.34 | |
LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
8193 | |
dodecan-1-ol |
C12H26O | |
MFCD00004753 | |
1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
CHEBI:28878 | |
CCCCCCCCCCCCO |
Specifications
112-53-8 | |
0.833g/mL at 25°C | |
121°C (249.8°F) | |
C12H26O | |
n20/D 1.442 | |
UN3077 | |
1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
CCCCCCCCCCCCO | |
186.34 | |
7.4 (vs air) | |
186.33g/mol | |
1-Dodecanol |
22°C to 26°C | |
260°C to 262°C | |
Conforms to structure | |
CH3(CH2)11OH | |
MFCD00004753 | |
1738860 | |
LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
dodecan-1-ol | |
8193 | |
CHEBI:28878 | |
≥98% |
Safety and Handling
P305 + P351 + P338
missing translation for 'dotInformation' : 9/PGIII
missing translation for 'rtecsNumber' : 203-982-0
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title