missing translation for 'onlineSavingsMsg'
Learn More

(1-Dodecyl)trimethylammonium bromide, 99%, Thermo Scientific Chemicals

47.64 € - 470.34 €

Chemical Identifiers

CAS 1119-94-4
Molecular Formula C15H34BrN
Molecular Weight (g/mol) 308.348
MDL Number MFCD00011767
InChI Key XJWSAJYUBXQQDR-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
PubChem CID 14249
ChEBI CHEBI:282662
IUPAC Name dodecyl(trimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Br-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469652
SDS View Documents
Thermo Scientific Alfa Aesar
A10761.09
10 g
47.64 €
10g
Please to purchase this item. Need a web account? Register with us today!
11479652
SDS View Documents
Thermo Scientific Alfa Aesar
A10761.18
50 g
151.76 €
50g
Please to purchase this item. Need a web account? Register with us today!
11489652
SDS View Documents
Thermo Scientific Alfa Aesar
A10761.30
250 g
470.34 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
Specifications

Chemical Identifiers

1119-94-4
308.348
XJWSAJYUBXQQDR-UHFFFAOYSA-M
14249
dodecyl(trimethyl)azanium;bromide
C15H34BrN
MFCD00011767
dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
CHEBI:282662
CCCCCCCCCCCC[N+](C)(C)C.[Br-]

Specifications

1119-94-4
C15H34BrN
UN3077
Hygroscopic
Soluble in water.
CCCCCCCCCCCC[N+](C)(C)C.[Br-]
308.348
CHEBI:282662
99%
∼245°C (decomposition)
MFCD00011767
3597463
dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
XJWSAJYUBXQQDR-UHFFFAOYSA-M
dodecyl(trimethyl)azanium;bromide
14249
308.35
(1-Dodecyl)trimethylammonium bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 214-290-3

missing translation for 'rtecsNumber' : BQ3195000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.