Learn More
1-Dodecylamine, 97%, Thermo Scientific Chemicals
27.44 € - 164.82 €
Chemical Identifiers
CAS | 124-22-1 |
---|---|
Molecular Formula | C12H27N |
Molecular Weight (g/mol) | 185.36 |
MDL Number | MFCD00008154 |
InChI Key | JRBPAEWTRLWTQC-UHFFFAOYSA-N |
Synonym | dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 |
PubChem CID | 13583 |
IUPAC Name | dodecan-1-amine |
SMILES | CCCCCCCCCCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11462917
|
Thermo Scientific Alfa Aesar
A15515.22 |
100 g |
27.44 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11472917
|
Thermo Scientific Alfa Aesar
A15515.36 |
500 g |
98.89 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468750
|
Thermo Scientific Alfa Aesar
A15515.A1 |
1 kg |
164.82 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.
Solubility
Soluble in water (78 mg/L 25°C).
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Chemical Identifiers
124-22-1 | |
185.36 | |
JRBPAEWTRLWTQC-UHFFFAOYSA-N | |
13583 | |
CCCCCCCCCCCCN |
C12H27N | |
MFCD00008154 | |
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 | |
dodecan-1-amine |
Specifications
124-22-1 | |
0.802 | |
115°C (239°F) | |
C12H27N | |
UN3259 | |
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 | |
JRBPAEWTRLWTQC-UHFFFAOYSA-N | |
dodecan-1-amine | |
13583 | |
≥98% |
27°C to 31°C | |
64°C to 66°C (0.05 mm) | |
≥98% | |
MFCD00008154 | |
1633576 | |
Soluble in water (78mg/L 25°C). | |
CCCCCCCCCCCCN | |
185.36 | |
185.36 | |
1-Dodecylamine |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H304-H314-H335-H373
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 204-690-6
missing translation for 'rtecsNumber' : JR6475000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.