missing translation for 'onlineSavingsMsg'
Learn More
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate, 98%, Thermo Scientific Chemicals Product Code.: 11453857

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Product Code. 11453857
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11453857

Brand: Thermo Scientific Alfa Aesar A18305.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52186-89-7
Molecular Formula C24H24BF4O2P
Molecular Weight (g/mol) 462.23
MDL Number MFCD00051879
InChI Key RGJYRMUGSAFITK-UHFFFAOYSA-N
Synonym 1-carbethoxycyclopropyltriphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl triphenylphosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonyl cyclopropyl triphenylphosphanium tetrafluoroborate, acmc-20aowe, 1-carboethoxycyclopropyl triphenylphosphonium tetrafluoroborate, 1-carbethoxycyclopropyl tris phenyl phosphonium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphanium tetrafluoroborate, 1-ethoxycarbonylcyclopropyl-triphenyl-phosphonium tetrafluoroborateShow MoreShow Less
PubChem CID 11016017
SMILES F[B-](F)(F)F.CCOC(=O)C1(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 173°C to 179°C
Quantity 1 g
Beilstein 4650673
Solubility Information Slightly soluble in water.
IUPAC Name [1-(ethoxycarbonyl)cyclopropyl]triphenylphosphanium; tetrafluoroboranuide
Formula Weight 462.23
Percent Purity 98%
Chemical Name or Material (1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10185617 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18305.03,A18305.14,A18305.06
Lot NumberJ1490A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18305.03,A18305.14,A18305.06
Lot Number10229612 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18305.03,A18305.14,A18305.06
Lot Number10183247 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18305.03,A18305.14,A18305.06
Lot Number10141987 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18305.03,A18305.14,A18305.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1-Ethoxycarbonylcyclopropyl)triphenylphosphonium tetrafluoroborate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.