Learn More
1-Ethylpiperidine, 99%, Thermo Scientific Chemicals
75.38 € - 309.54 €
Chemical Identifiers
CAS | 766-09-6 |
---|---|
Molecular Formula | C7H15N |
Molecular Weight (g/mol) | 113.2 |
MDL Number | MFCD00006507 |
InChI Key | HTLZVHNRZJPSMI-UHFFFAOYSA-N |
Synonym | n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene |
PubChem CID | 13007 |
ChEBI | CHEBI:39017 |
IUPAC Name | 1-ethylpiperidine |
SMILES | CCN1CCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10132530
|
Thermo Scientific Acros
118885000 |
500 mL | Glass bottle |
75.38 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10463055
|
Thermo Scientific Acros
118880025 |
2.5 L | Glass bottle |
309.54 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
766-09-6 | |
113.2 | |
HTLZVHNRZJPSMI-UHFFFAOYSA-N | |
13007 | |
1-ethylpiperidine |
C7H15N | |
MFCD00006507 | |
n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene | |
CHEBI:39017 | |
CCN1CCCCC1 |
Specifications
766-09-6 | |
100.0 | |
Colorless to Yellow | |
129.0°C to 131.0°C | |
Authentic | |
C7H15N | |
MFCD00006507 | |
0.824 | |
Solubility in water: 50g/L (20°C) | |
CCN1CCCCC1 | |
113.2 | |
CHEBI:39017 | |
99% | |
1-Ethylpiperidine, 99% |
98.5 | |
-20.0°C | |
0.8240g/mL | |
17°C | |
98.5% min. (GC) | |
1.4430 to 1.4450 | |
20,17 | |
n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene | |
HTLZVHNRZJPSMI-UHFFFAOYSA-N | |
1-ethylpiperidine | |
13007 | |
113.2 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-161-6
missing translation for 'rtecsNumber' : TN0250000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.