missing translation for 'onlineSavingsMsg'
Learn More
1-Ethylpiperidine, 99%, Thermo Scientific Chemicals Product Code.: 15354968

1-Ethylpiperidine, 99%, Thermo Scientific Chemicals

Product Code. 15354968
10 L
Click to view available options
Quantity:
10 L
This item is not returnable. View return policy

Product Code. 15354968

Brand: Thermo Scientific Acros 118880100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 766-09-6
Melting Point -20.0°C
Density 0.8240g/mL
Boiling Point 129.0°C to 131.0°C
Flash Point 17°C
Assay Percent Range 98.5% min. (GC)
Packaging Plastic Drum
Molecular Formula C7H15N
Refractive Index 1.4430 to 1.4450
Quantity 10 L
Beilstein 20,17
Specific Gravity 0.824
Synonym n-ethylpiperidine, piperidine, 1-ethyl, n-aethylpiperidin, n-ethyl piperidine, ethylpiperidine, n-aethylpiperidin german, 1-ethyl-piperidine, n-ethyl-piperidine, 1-ethyl piperidine, 1-ethylpiperidene
Solubility Information Solubility in water: 50g/L (20°C).
InChI Key HTLZVHNRZJPSMI-UHFFFAOYSA-N
SMILES CCN1CCCCC1
IUPAC Name 1-ethylpiperidine
Molecular Weight (g/mol) 113.2
PubChem CID 13007
ChEBI CHEBI:39017
Formula Weight 113.2
Percent Purity 99%
Chemical Name or Material 1-Ethylpiperidine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0352795 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number118880025,118880100,118885000
Lot NumberA0419729 Certificate TypeCertificate of Analysis Date05/12/2022 Catalog Number118880025,118880100,118885000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Ethylpiperidine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.