missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylquinolinium iodide, 98%, Thermo Scientific™

Product Code. 11413458
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-35-5
Molecular Formula C11H12IN
Molecular Weight (g/mol) 285.128
MDL Number MFCD00041996
InChI Key PMYUGMDDIBOXQM-UHFFFAOYSA-M
Synonym 1-ethylquinolinium iodide, quinoline ethiodide, 1-ethylquinolin-1-ium iodide, n-ethylquinolinium iodide, quinolinium, 1-ethyl-, iodide, quinolinium, 1-ethyl-, iodide 1:1, 1-ethylquinoliniumiodide, acmc-1b72m, quinoline ethyl iodide
PubChem CID 69446
IUPAC Name 1-ethylquinolin-1-ium;iodide
SMILES CC[N+]1=CC=CC2=CC=CC=C21.[I-]

Specifications

Melting Point 156°C to 159°C
Quantity 100 g
Sensitivity Light sensitive
Formula Weight 285.13
Percent Purity 98%
Chemical Name or Material 1-Ethylquinolinium iodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'