missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3-nitrobenzene, 98+%

Product Code. 11447236
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11447236

Brand: Thermo Scientific Alfa Aesar B25553.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 402-67-5 | C6H4FNO2 | 141.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-67-5
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD00007196
InChI Key WMASLRCNNKMRFP-UHFFFAOYSA-N
Synonym 3-fluoronitrobenzene, m-fluoronitrobenzene, m-nitrofluorobenzene, benzene, 1-fluoro-3-nitro, 3-fluoro-1-nitrobenzene, 1-fluoro-3-nitro-benzene, 3-fluoro-nitrobenzene, 3-nitro-fluorobenzene, pubchem2286, 3-fluoro nitro benzene
PubChem CID 9823
IUPAC Name 1-fluoro-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(F)=C1

Specifications

Melting Point 2°C
Density 1.325
Boiling Point 204°C to 206°C
Flash Point 76°C (168°F)
Refractive Index 1.525
Quantity 25 g
UN Number UN2810
Beilstein 1862210
Formula Weight 141.1
Percent Purity ≥98%
Chemical Name or Material 1-Fluoro-3-nitrobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.