missing translation for 'onlineSavingsMsg'
Learn More
1-Fluoronaphthalene, 98%, Thermo Scientific Chemicals Product Code.: 11428307

1-Fluoronaphthalene, 98%, Thermo Scientific Chemicals

Product Code. 11428307
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11428307

Brand: Thermo Scientific Alfa Aesar A18378.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoronaphthalene is used in t-BuLi-mediated synthesis of 6-substituted phenanthridines. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-38-0
Molecular Formula C10H7F
Molecular Weight (g/mol) 146.164
MDL Number MFCD00003873
InChI Key CWLKTJOTWITYSI-UHFFFAOYSA-N
Synonym fluoronaphthalene, naphthalene, 1-fluoro, alpha-fluoronaphthalene, 1-fluornaftalen, .alpha.-fluoronaphthalene, 1-fluornaftalen czech, 1-fluoro naphthalene, fluoronaphtalene, fluoronapthalene, 1-fluoronapthalene
PubChem CID 9450
IUPAC Name 1-fluoronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2F

Specifications

Melting Point -10°C
Density 1.331
Boiling Point 215°C to 217°C
Flash Point 65°C (149°F)
Refractive Index 1.593
Quantity 25 g
Beilstein 1906413
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 146.17
Percent Purity 98%
Chemical Name or Material 1-Fluoronaphthalene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Fluoronaphthalene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.