missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals

Product Code. 11904571
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207300-90-1
InChI Key XWZCREJRXRKIRQ-UHFFFAOYSA-M
Synonym sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
PubChem CID 23687711
IUPAC Name sodium;heptane-1-sulfonate;hydrate
SMILES CCCCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Absorbance (2.7g in 100mL water) (pH 3), (2.7 g in 100 ml water) (pH 3) at 0nm, 0.06 max. at 254nm, 0.06 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm
Melting Point >300.0°C
Color White
Loss on Drying 7.4% to 9% (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C7H15NaO3S·H2O
Linear Formula CH3(CH2)6SO3Na·H2O
Quantity 500 g
Solubility Information (10% w/v in water) Clear colorless
Molecular Weight (g/mol) 220.27
Formula Weight 220.27
Physical Form Powder or Flakes
Chemical Name or Material 1-Heptanesulfonic acid sodium salt monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.