missing translation for 'onlineSavingsMsg'
Learn More

1-Heptanol, 98%, Thermo Scientific Chemicals

Product Code. 10409420
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00002986
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
PubChem CID 8129
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Acidity 0.5mg KOH/g max.
CAS Min % 97.5
CAS Max % 100.0
Melting Point -35.0°C
Density 0.8219g/mL
Boiling Point 176.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)6OH
Refractive Index 1.4230 to 1.4250
Quantity 250 mL
Beilstein 01, 414
Residue after Evaporation 0.01% max.
Merck Index 15, 4696
Specific Gravity 0.8219
Solubility Information Solubility in water: 2.85g/L (100°C). Other solubilities: 5.15g/L water (130°C), soluble in ethanol, ether and usual organic, solvents
Formula Weight 116.2
Percent Purity 98%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 1-Heptanol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Heptanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.