missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexane Sulfonic Acid, Sodium Salt, 98+%, Ion Pair Chromatography, Certified, Fisher Chemical™
425.77 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
1-Hexane Sulfonic Acid, Sodium Salt, 98+%, Ion Pair Chromatography, Certified, Fisher Chemical, Quantity: 100g, CAS: 2832-45-3, White, Melting Point: 300 deg.C, Molecular Formula: C6H13NaO3S, Molecular Weight: 188.217, Packaging: Plastic powder jar
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 300°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
IUPAC Name | sodium;hexane-1-sulfonate |
Formula Weight | 188.22g/mol |
Grade | Chromatography |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'