Learn More
1-Hexanol, 99%, Thermo Scientific Chemicals
26.83 € - 84.22 €
Chemical Identifiers
CAS | 111-27-3 |
---|---|
Molecular Formula | C6H14O |
Molecular Weight (g/mol) | 102.177 |
MDL Number | MFCD00002982 |
InChI Key | ZSIAUFGUXNUGDI-UHFFFAOYSA-N |
Synonym | 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinolShow More |
PubChem CID | 8103 |
ChEBI | CHEBI:87393 |
IUPAC Name | hexan-1-ol |
SMILES | CCCCCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493757
|
Thermo Scientific Alfa Aesar
A18232.AP |
500 mL |
26.83 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403767
|
Thermo Scientific Alfa Aesar
A18232.0F |
2500 mL |
84.22 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Hexanol is used as a precursor to plasticizers, chemical intermediate for pharmaceuticals, perfume esters and antiseptics. Further, it serves as a perturbing agent on actomyosin adenosine triphosphatease. In addition to this, it is used to modulate the function of actomyosin motor.
Solubility
Miscible with ethanol, acetone, chloroform, ether, benzene. Slightly miscible with carbon tetrachloride and water.
Notes
Incompatible with strong acids, acid chlorides, acid anhydrides and strong oxidizing agents.
Chemical Identifiers
111-27-3 | |
102.177 | |
ZSIAUFGUXNUGDI-UHFFFAOYSA-N | |
8103 | |
hexan-1-ol |
C6H14O | |
MFCD00002982 | |
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinolShow More | |
CHEBI:87393 | |
CCCCCCO |
Specifications
111-27-3 | |
0.814 g/mL | |
60°C (140°F) | |
C6H14O | |
1.418 | |
UN2282 | |
14,4697 | |
Miscible with ethanol,acetone,chloroform,ether,benzene. Slightly miscible with carbon tetrachloride and water. | |
CCCCCCO | |
102.177 | |
CHEBI:87393 | |
99% |
°C to 52°C | |
157°C | |
99% | |
CH3(CH2)5OH | |
MFCD00002982 | |
969167 | |
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol | |
ZSIAUFGUXNUGDI-UHFFFAOYSA-N | |
hexan-1-ol | |
8103 | |
102.18 | |
1-Hexanol |
Safety and Handling
GHS H Statement
H226-H302
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P362-P370+P378q-P374-P380-P501c
H226-H302+H312-H319
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HEXANOLS
missing translation for 'einecsNumber' : 203-852-3
missing translation for 'rtecsNumber' : MQ4025000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- 1-Hexanol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : 1-hexanol
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.