missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Indanone, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Description
1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.
Solubility
Soluble in water (6.5 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
1-Indanone is an oxidation product of indan, a component of fuels, solvents and varnishes. 1-Indanone is also a metabolite of Thalidomide that has been shown to inhibit the attachment of tumor cells to concanavalin A coated plastic surfaces.
Solubility
Soluble in water (6.5 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 38°C to 42°C |
Density | 1.103 |
Boiling Point | 243°C to 245°C |
Flash Point | 113°C (235°F) |
Quantity | 25 g |
Beilstein | 507957 |
Solubility Information | Soluble in water (6.5g/L at 20°C). |
Formula Weight | 132.16 |
Percent Purity | ≥99% |
Chemical Name or Material | 1-Indanone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'