missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-isopropylbenzene, 95%, Thermo Scientific Chemicals

Product Code. 15428799
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 19099-54-8
Density 1.54
Boiling Point 110°C (20 mmHg)
Odor Aromatic
Molecular Formula C9H11I
Refractive Index 1.583
MDL Number MFCD00013704
Quantity 5 g
Beilstein 2351872
Sensitivity Light sensitive
Synonym 1-iodo-2-isopropylbenzene, 2-iodoisopropylbenzene, 2-iodocumene, o-iodocumene, 1-iodo-2-isopropyl-benzene, benzene, 1-iodo-2-1-methylethyl, o-i-propyliodobenzene, 2-isopropyliodobenzene, 2-iodoisopropyl benzene, pubchem12584
InChI Key SORQIYFSJAWBNQ-UHFFFAOYSA-N
SMILES CC(C)C1=CC=CC=C1I
IUPAC Name 1-iodo-2-propan-2-ylbenzene
Molecular Weight (g/mol) 246.09
PubChem CID 87931
Formula Weight 246.09
Percent Purity 95%
Chemical Name or Material 1-Iodo-2-isopropylbenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.