Learn More
1-Iododecane, 98%, stab. with copper, Thermo Scientific Chemicals
Description
1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Iododecane is used as a precursor to prepare 9-nonadecanone through palladium-catalyzed carbonylative cross-coupling reaction with 9-octyl-9-borabicyclo[3.3.l]nonane. It is also used as an intermediate in organic synthesis.
Solubility
Immiscible with water.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases. Keep the container tightly closed.

Specifications
Specifications
Melting Point | -16°C |
Density | 1.257 |
Boiling Point | 264°C |
Flash Point | 96°C (204°F) |
Linear Formula | CH3(CH2)9I |
Refractive Index | 1.485 |
Quantity | 50 g |
Beilstein | 1735228 |
Sensitivity | Light sensitive |
Synonym | decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10222505 | Certificate of Analysis | 30/08/2024 | A16173.30,A16173.18,A16173.14 |
10148652 | Certificate of Analysis | 23/11/2021 | A16173.30,A16173.18,A16173.14 |
10199157 | Certificate of Analysis | 23/11/2021 | A16173.30,A16173.18,A16173.14 |
10166867 | Certificate of Analysis | 23/11/2021 | A16173.30,A16173.18,A16173.14 |
10222505 | Certificate of Analysis | 23/11/2021 | A16173.30,A16173.18,A16173.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.