missing translation for 'onlineSavingsMsg'
Learn More
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals Product Code.: 10215091

1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals

Product Code. 10215091
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10215091

Brand: Thermo Scientific Acros 330361000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2050-77-3
Molecular Formula C10H21I
Molecular Weight (g/mol) 268.18
MDL Number MFCD00001085
InChI Key SKIDNYUZJPMKFC-UHFFFAOYSA-N
PubChem CID 16314
IUPAC Name 1-iododecane
SMILES CCCCCCCCCCI

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.2500g/mL
Boiling Point 124.0°C (11.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)9I
Refractive Index 1.4845 to 1.4865
Quantity 100 g
Beilstein 01,168
Specific Gravity 1.25
Synonym decyl iodide, decane, 1-iodo, n-decyl iodide, iododecane, 1-iodanyldecane, 1-decyl iodide, 1-iodo-decane, 1-iod0decane, decane,1-iodo, pubchem3942
Stabilizer copper
Formula Weight 268.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Iododecane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0417079 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number330361000,330365000,330360250
Lot NumberA0463984 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog Number330361000,330365000,330360250
Lot NumberA0377402 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number330361000,330365000,330360250
Lot NumberA0417079 Certificate TypeCertificate of Analysis Date30/08/2021 Catalog Number
Lot NumberA0415359 Certificate TypeCertificate of Analysis Date29/06/2021 Catalog Number330361000,330365000,330360250
compliance-icons
Product Identifier
  • 1-Iododecane, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.