Learn More
1-Iodooctane, 98+%, stab. with copper, Thermo Scientific Chemicals
Description
Alkylation of [2,2?-([2,2?-bipyridine]-6,6?-diyl)bis[phenolato]-N,N?,O,O?]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation of [2,2′-([2,2′-bipyridine]-6,6′-diyl)bis[phenolato]-N,N′,O,O′]nickel(II)was performed using the catalytic reduction of 1-iodooctane. Primary alkyl iodide 1-iodooctane reacts with lithium aluminum deuteride (LAD) in a 1:0.2 ratio to form 1-deuteriooctane.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, bases.

Specifications
Specifications
Melting Point | -46°C |
Density | 1.325 |
Boiling Point | 225°C to 226°C |
Flash Point | 95°C (203°F) |
Linear Formula | CH3(CH2)7I |
Refractive Index | 1.4878 |
Quantity | 100 g |
Beilstein | 1697479 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10241044 | Certificate of Analysis | 10/06/2022 | B22214.14,B22214.36,B22214.22 |
10240114 | Certificate of Analysis | 12/04/2022 | B22214.14,B22214.36,B22214.22 |
10200992 | Certificate of Analysis | 23/11/2021 | B22214.14,B22214.36,B22214.22 |
10176825 | Certificate of Analysis | 23/11/2021 | B22214.14,B22214.36,B22214.22 |
10216911 | Certificate of Analysis | 23/11/2021 | B22214.14,B22214.36,B22214.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.