missing translation for 'onlineSavingsMsg'
Learn More

1-Iodopropane, 98+%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11476987
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Iodopropane is used in wide range of medicals industrial applications as well as in human and animal nutrition products, pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents etc. It is also used as a solvent and intermediate of organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodopropane is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals. Iodine derivatives are also used as organic building blocks, analytical reagents etc. It is also used as a solvent and intermediate of organic synthesis.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-08-4
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001095
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym propyl iodide, n-propyl iodide, propane, 1-iodo, propane, iodo, 1-iodo-propane, 1-jodpropan, iodopropane, unii-nd3629ke2k, propyliodide, iodopropanes
PubChem CID 33643
SMILES CC(C)I

Specifications

Melting Point °C to 101°C
Density 1.744
Boiling Point 101°C to 102°C
Flash Point 44°C (111°F)
Assay Percent Range ≥98%, Stabilized with copper
Linear Formula CH3CH2CH2I
Refractive Index 1.505
Quantity 100 g
UN Number UN2392
Beilstein 505937
Sensitivity Light sensitive
Merck Index 14,7862
Solubility Information Fully miscible in water.
IUPAC Name 1-iodopropane
Formula Weight 169.99
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 1-Iodopropane, stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'