Learn More
Wheaton™ 1 μL Inoculation Loop
Inoculation loop calibrated to deliver a known volume of fluid sample to enable accurate microbiological analysis.
Brand:  Wheaton™ 711310
 View more versions of this product
Product Code. 16435009
  • 365.00 € / Pack of 6000
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Meets quantitative standards for management of microbiological samples
  • For accurate liquid transfer and plating
  • Calibrated
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single-use
199 mm
Sterile
Blue
Packaged in peel pouch
Not Autoclavable
1 μL
ABS
Standard
6000
Yes