missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride, 90%, Thermo Scientific™

46.70 € - 113.00 €

Chemical Identifiers

CAS 906352-57-6
Molecular Formula C9H8Cl2N2O
Molecular Weight (g/mol) 231.08
MDL Number MFCD09817460
InChI Key SGNORNZSRSQMEE-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzimidazole-5-carbonyl chloride hydrochloride, 1-methyl-1,3-benzodiazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzoimidazole-5-carbonyl chloride.hcl, 1-methylbenzimidazole-5-carbonyl chloride, chloride, 1-methyl-1h-benzo d imidazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzimidazole-5-carbonyl chloride-hydrogen chloride 1/1, 1h-benzimidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1
PubChem CID 24229458
IUPAC Name 1-methylbenzimidazole-5-carbonyl chloride;hydrochloride
SMILES Cl.CN1C=NC2=CC(=CC=C12)C(Cl)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11310577
View Documents
Thermo Scientific Maybridge
CC13652CB
250 mg Amber Glass Bottle
46.70 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10706295
View Documents
Thermo Scientific Maybridge
CC13652DA
1 g Amber Glass Bottle
113.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

906352-57-6
231.08
SGNORNZSRSQMEE-UHFFFAOYSA-N
24229458
Cl.CN1C=NC2=CC(=CC=C12)C(Cl)=O
C9H8Cl2N2O
MFCD09817460
1-methyl-1h-benzimidazole-5-carbonyl chloride hydrochloride, 1-methyl-1,3-benzodiazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzoimidazole-5-carbonyl chloride.hcl, 1-methylbenzimidazole-5-carbonyl chloride, chloride, 1-methyl-1h-benzo d imidazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzimidazole-5-carbonyl chloride-hydrogen chloride 1/1, 1h-benzimidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1
1-methylbenzimidazole-5-carbonyl chloride;hydrochloride

Specifications

906352-57-6
100.0
C9H8Cl2N2O
1-methyl-1h-benzimidazole-5-carbonyl chloride hydrochloride, 1-methyl-1,3-benzodiazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzoimidazole-5-carbonyl chloride.hcl, 1-methylbenzimidazole-5-carbonyl chloride, chloride, 1-methyl-1h-benzo d imidazole-5-carbonyl chloride hydrochloride, 1-methyl-1h-benzimidazole-5-carbonyl chloride-hydrogen chloride 1/1, 1h-benzimidazole-5-carbonylchloride, 1-methyl-, hydrochloride 1:1
Cl.CN1C=NC2=CC(=CC=C12)C(Cl)=O
231.08
231.08
1-Methyl-1H-benzimidazole-5-carbonyl chloride hydrochloride
89.5
90%
MFCD09817460
SGNORNZSRSQMEE-UHFFFAOYSA-N
1-methylbenzimidazole-5-carbonyl chloride;hydrochloride
24229458
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications