missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-3-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10746905
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 25016-20-0
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00464254
InChI Key YBFIKNNFQIBIQZ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid
PubChem CID 573176
ChEBI CHEBI:74738
IUPAC Name 1-methylpyrazole-3-carboxylic acid
SMILES CN1C=CC(=N1)C(O)=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 126.11
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-pyrazole-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.