missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%

Product Code. 15400988
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15400988

Brand: Thermo Scientific Alfa Aesar H54785.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 288148-34-5 | C4H5ClN2O2S | 180.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288148-34-5
Molecular Formula C4H5ClN2O2S
Molecular Weight (g/mol) 180.606
MDL Number MFCD04968667
InChI Key RDAVKKQKMLINOH-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl, chloro 1-methylpyrazol-4-yl sulfone, 4-chlorosulphonyl-1-methyl-1h-pyrazole, 1-methylpyrazole-4-sulfonylchloride, 1-methyl-4-pyrazolesulfonyl chloride, 1h-pyrazole-4-sulfonylchloride, 1-methyl, 1h-pyrazole-4-sulfonylchloride,1-methyl-9ci, 4-chlorosulfonyl-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazole-4-sulphonyl chloride
PubChem CID 17024730
IUPAC Name 1-methylpyrazole-4-sulfonyl chloride
SMILES CN1C=C(C=N1)S(=O)(=O)Cl

Specifications

Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 179.98
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-4-sulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.