missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid, 97%, Thermo Scientific™ Product Code.: 10453024

1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10453024
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10453024

Brand: Thermo Scientific Maybridge CC70901CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 116855-09-5
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.163
MDL Number MFCD11109332
InChI Key WEZYUZRHWLFJDN-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazolo 3,4-b pyridine-3-carboxylic acid, 1-methylpyrazolo 3,4-b pyridine-3-carboxylic acid, 1h-pyrazolo 3,4-b pyridine-3-carboxylicacid,1-methyl, 1h-pyrazolo 3,4-b pyridine-3-carboxylicacid, 1-methyl, 1h-pyrazolo 3,4-b pyridine-3-carboxylic acid, 1-methyl, acmc-1bnof, 1-methyl-3-pyrazolo 3,4-b pyridinecarboxylic acid, 1-methylpyrazolo 5,4-b pyridine-3-carboxylic acidShow MoreShow Less
PubChem CID 14140175
IUPAC Name 1-methylpyrazolo[3,4-b]pyridine-3-carboxylic acid
SMILES CN1C2=C(C=CC=N2)C(=N1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-pyrazolo[3, 4-b]pyridine-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.