missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals Product Code.: 13237069

1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals

Product Code. 13237069
5 L
Click to view available options
Quantity:
5 L
25 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 13237069

Brand: Thermo Scientific Acros 449180050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24.0°C
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 5 L
Flash Point 91°C
Packaging Metal Drum
Refractive Index 1.4690 to 1.4710
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.03
Viscosity 1.67 mPa.s (20°C)
Merck Index 15,619
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Percent Purity 98%
Grade Pure
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457335 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number449180050,449180250
Lot NumberA0466045 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog Number449180050,449180250
Lot NumberA0460803 Certificate TypeCertificate of Analysis Date01/03/2024 Catalog Number449180050,449180250
Lot NumberA0457335 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number449180050,449180250
Lot NumberA0451123 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number449180050,449180250
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Methyl-2-pyrrolidone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.