Learn More
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
34.60 € - 291.00 €
Chemical Identifiers
CAS | 872-50-4 |
---|---|
Molecular Formula | C5H9NO |
Molecular Weight (g/mol) | 99.133 |
MDL Number | MFCD00003193 |
InChI Key | SECXISVLQFMRJM-UHFFFAOYSA-N |
Synonym | 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl |
PubChem CID | 13387 |
ChEBI | CHEBI:7307 |
IUPAC Name | 1-methylpyrrolidin-2-one |
SMILES | CN1CCCC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10383692
|
Thermo Scientific Acros
364381000 |
100 mL | AcroSeal™ Glass Bottle |
34.60 €
100mL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents. Find out more about AcroSeal™ packaging. |
|||||||||
10079051
|
Thermo Scientific Acros
364380010 |
1 L | AcroSeal™ Glass Bottle |
157.00 €
1 L |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents. Find out more about AcroSeal™ packaging. |
|||||||||
10304122
|
Thermo Scientific Acros
364380025 |
2.5 L | AcroSeal™ Glass Bottle |
291.00 €
2.50 L |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents. Find out more about AcroSeal™ packaging. |
|||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
872-50-4 | |
99.133 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
13387 | |
1-methylpyrrolidin-2-one |
C5H9NO | |
MFCD00003193 | |
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
CHEBI:7307 | |
CN1CCCC1=O |
Specifications
-24.0°C | |
99.4 | |
100.0 | |
202.0°C | |
Authentic | |
1.4690 to 1.4710 | |
1.03 | |
MFCD00003193 | |
21, 237 | |
01,696; 02,281; 09,316; 11,346 | |
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
1-methylpyrrolidin-2-one | |
13387 | |
99.13 | |
Extra Dry over Molecular Sieve |
872-50-4 | |
Colorless | |
1.0300g/mL | |
91°C | |
Liquid | |
99.5% | |
C5H9NO | |
1.67 mPa.s (20°C) | |
0.005% Max. (Coulometric) | |
15, 6190 | |
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents | |
CN1CCCC1=O | |
99.133 | |
CHEBI:7307 | |
99.5% | |
1-Methyl-2-pyrrolidinone, Over Molecular Sieves |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 212-828-1
missing translation for 'rtecsNumber' : UY5790000
missing translation for 'tsca' : TSCA
- 1-Methyl-2-pyrrolidinone
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H360D-May damage the unborn child.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 1-methyl-2-pyrrolidone
RUO – Research Use Only