missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

Product Code. 15234228
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 872-50-4 | C5H9NO | 99.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -24.0°C
CAS 872-50-4
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 500 mL
Flash Point 91°C
Infrared Spectrum Authentic
Refractive Index 1.4690 to 1.4710
Assay Percent Range 99.5% min. (GC)
Specific Gravity 1.03
Molecular Formula C5H9NO
Viscosity 1.67 mPa.s (20°C)
Beilstein 21,237
Water 0.005% max. (Coulometric)
Fieser 01,696; 02,281; 09,316; 11,346
Merck Index 15,619
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES CN1CCCC1=O
IUPAC Name 1-methylpyrrolidin-2-one
Molecular Weight (g/mol) 99.13
Formula Weight 99.13
Percent Purity 99.50%
Grade Extra Dry over Molecular Sieve
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H360D-May damage the unborn child.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.