Learn More
1-Methyl-2-pyrrolidinone, 99.5%, for HPLC, Thermo Scientific Chemicals
Shop All Thermo Scientific ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Melting Point | -24.0°C |
Absorbance | 0.01 max. at 340nm, 0.01 max. at 450nm, 0.1 max. at 320nm, 0.5 max. at 300nm, 1 max. at 285nm |
CAS Min % | 99.4 |
CAS Max % | 100.0 |
Density | 1.0300g/mL |
Boiling Point | 202.0°C |
Quantity | 100 mL |
Flash Point | 91°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0451922 | Certificate of Analysis | 31/01/2025 | 368450025,368451000,368450010 |
A0458085 | Certificate of Analysis | 25/04/2024 | 368450025,368451000,368450010 |
A0458084 | Certificate of Analysis | 20/03/2024 | 368450025,368451000,368450010 |
A0446186 | Certificate of Analysis | 09/11/2023 | 368450025,368451000,368450010 |
A0446811 | Certificate of Analysis | 25/09/2023 | 368450025,368451000,368450010 |
Safety and Handling


- 1-Methyl-2-pyrrolidinone
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H360D-May damage the unborn child.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.