missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 99.5%, for HPLC

Product Code. p-2689565 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2.5 L
This item is not returnable. View return policy

Product Code. 10478172

Brand: Thermo Scientific undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 872-50-4 | C5H9NO | 99.133 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point -24.0°C
Absorbance 0.01 max. at 340nm, 0.01 max. at 450nm, 0.1 max. at 320nm, 0.5 max. at 300nm, 1 max. at 285nm
CAS Min % 99.4
CAS Max % 100.0
Density 1.0300g/mL
Boiling Point 202.0°C
Quantity 100 mL
Flash Point 91°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4690 to 1.4710
Assay Percent Range 99.5%
Specific Gravity 1.03
Viscosity 1.67 mPa.s (20°C)
Beilstein 21, 237
Water 0.05% max. (K.F.)
Fieser 01,696; 02,281; 09,316; 11,346
Merck Index 15, 6190
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 99.13
Percent Purity 99.5%
Grade HPLC
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.