missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
Melting Point | -24.0°C |
Density | 1.0300g/mL |
Boiling Point | 202.0°C |
Quantity | 10 L |
Flash Point | 91°C |
Packaging | Metal Drum |
Refractive Index | 1.4690 to 1.4710 |
Assay Percent Range | 99.4% min. (GC) |
Specific Gravity | 1.03 |
Viscosity | 1.67 mPa.s (20°C) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'