Learn More
1-Methyl-2-pyrrolidinone, 99%, extra pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 99%, C5H9NO, CAS Number-872-50-4, 1-methyl-2-pyrrolidinone, 1-methyl-2-pyrrolidone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidinone, n-methylpyrrolidone, 2.5L
109.55 € - 510.54 €
Chemical Identifiers
CAS | 872-50-4 |
---|---|
Molecular Formula | C5H9NO |
Molecular Weight (g/mol) | 99.133 |
MDL Number | MFCD00003193 |
InChI Key | SECXISVLQFMRJM-UHFFFAOYSA-N |
Synonym | 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methylShow More |
PubChem CID | 13387 |
ChEBI | CHEBI:7307 |
IUPAC Name | 1-methylpyrrolidin-2-one |
SMILES | CN1CCCC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10773891
|
Thermo Scientific Acros
127630025 |
2.5 L | Glass Bottle |
109.55 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023113
|
Thermo Scientific Acros
127630250 |
25 L | Metal Drum |
510.54 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
872-50-4 | |
99.133 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
13387 | |
1-methylpyrrolidin-2-one |
C5H9NO | |
MFCD00003193 | |
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methylShow More | |
CHEBI:7307 | |
CN1CCCC1=O |
Specifications
-24.0°C | |
98.5 | |
1.0300g/mL | |
91°C | |
Liquid | |
99% | |
C5H9NO | |
Vapor Pressure: 0.7mbar at 25°C | |
21, 237 | |
01,696; 02,281; 09,316; 11,346 | |
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
1-methylpyrrolidin-2-one | |
13387 | |
99.13 | |
Extra Pure |
872-50-4 | |
100.0 | |
202.0°C | |
Authentic | |
1.4690 to 1.4710 | |
1.03 | |
MFCD00003193 | |
1.67 mPa.s (20°C) | |
0.05% max. (K.F.) | |
15, 6190 | |
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents | |
CN1CCCC1=O | |
99.133 | |
CHEBI:7307 | |
99% | |
1-Methyl-2-pyrrolidinone |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- 1-Methyl-2-pyrrolidinone
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H360D-May damage the unborn child.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 1-methyl-2-pyrrolidone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.