missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, 99+%, for spectroscopy, Thermo Scientific Chemicals

Product Code. 10452221
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Absorbance (1 cm cell vs water), 0.01 at 340nm, 0.04 at 320nm, 0.1 at 300nm, 0.55 at 270nm, 1 at 262nm
Density 1.0300g/mL
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99+%
Vapor Pressure Vapor Pressure: 0.7mbar at 25°C
Beilstein 21, 237
Water 0.05% max. (K.F.)
Fieser 01,696; 02,281; 09,316; 11,346
Merck Index 15, 6190
Formula Weight 99.13
Percent Purity ≥99.0%
Grade Spectroscopy
Chemical Name or Material 1-Methyl-2-pyrrolidinone, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'