missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Spécification
Spécification
Melting Point | -24.0°C |
Density | 1.0300g/mL |
Boiling Point | 202.0°C |
Quantity | 10 L |
Flash Point | 91°C |
Infrared Spectrum | Authentic |
Packaging | Plastic Drum |
Physical Form | Liquid |
Refractive Index | 1.4690 to 1.4710 |
Specific Gravity | 1.03 |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'