missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-pyrrolidinone, CHROMASOLV™ Plus, for HPLC, ≥99%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
Packaging:
Glass Bottle
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Specifications
Specifications
Melting Point | -24°C (literature) |
Absorbance | max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.10 at 325 nm, max. 0.50 at 300 nm, max. 1.0 at 285 nm |
CAS Max % | 100.0000% |
Density | 1.028 g/mL at 25 °C(lit.) |
Boiling Point | 204°C |
Quantity | 4 x 4 L |
Flash Point | 91°C |
Packaging | Glass Bottle |
Linear Formula | C5H9NO |
Beilstein | 106420 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction