missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol, 97%, Thermo Scientific™

Product Code. 10386664
Amber Glass Bottle; 1g
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 879896-49-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H10N2OS
MDL Number MFCD08435885
Quantity 1g
Synonym 1-methyl-3-thien-2-yl-1h-pyrazol-5-yl methanol, 1h-pyrazole-5-methanol,1-methyl-3-2-thienyl, 2-methyl-5-thiophen-2-yl pyrazol-3-yl methanol, 1-methyl-3-thiophen-2-yl-1h-pyrazol-5-yl methanol, 1-methyl-3-2-thienyl pyrazol-5-yl methan-1-ol
InChI Key CHKYTDUITSTBNJ-UHFFFAOYSA-N
SMILES CN1C(=CC(=N1)C2=CC=CS2)CO
IUPAC Name (2-methyl-5-thiophen-2-ylpyrazol-3-yl)methanol
Molecular Weight (g/mol) 194.252
PubChem CID 18525810
Formula Weight 194.26
Percent Purity 97%
Chemical Name or Material (1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1-Methyl-3-thien-2-yl-1H-pyrazol-5-yl)methanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.