missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-thien-2-yl-1H-pyrazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10739405
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-48-7
Molecular Formula C9H8N2OS
Molecular Weight (g/mol) 192.24
MDL Number MFCD08435884
InChI Key RKARRIJZFBGKSH-UHFFFAOYSA-N
Synonym 1-methyl-3-thien-2-yl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-thiophen-2-yl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-thien-2-yl-1h-pyrazole-5-carboxaldehyde, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-2-thienyl, 2-methyl-5-thiophen-2-yl pyrazole-3-carbaldehyde, 1-methyl-3-2-thienyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-2-thienyl pyrazole-5-carbaldehyde, 1-methyl-3-2-thienyl-pyrazole-5-carbaldehyde, 2-5-formyl-1-methyl-1h-pyrazol-3-yl thiophene
PubChem CID 18525809
IUPAC Name 2-methyl-5-thiophen-2-ylpyrazole-3-carbaldehyde
SMILES CN1N=C(C=C1C=O)C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 192.24
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-thien-2-yl-1H-pyrazole-5-carbaldehyde
compliance-icons
Product Identifier
  • 1-Methyl-3-thien-2-yl-1H-pyrazole-5-carbaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Methyl-3-thien-2-yl-1H-pyrazole-5-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.