missing translation for 'onlineSavingsMsg'
Learn More
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Thermo Scientific™ Product Code.: 11366815

1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Thermo Scientific™

Product Code. 11366815
5 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 11366815

Brand: Thermo Scientific Maybridge AW00087DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 519056-67-8
Molecular Formula C5H4ClF3N2O2S
Molecular Weight (g/mol) 248.604
InChI Key HJFWSNIHZZKYIK-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride,1-methyl-3-trifluoromethyl, chloro 1-methyl-3-trifluoromethyl pyrazol-4-yl sulfone, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonylchloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethylShow MoreShow Less
PubChem CID 2794561
IUPAC Name 1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride
SMILES CN1C=C(C(=N1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.