missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Methyl-4-piperidnyl)methanamine, 97%, Thermo Scientific™
192.00 € - 755.00 €
Chemical Identifiers
CAS | 7149-42-0 |
---|---|
Molecular Formula | C7H16N2 |
Molecular Weight (g/mol) | 128.22 |
MDL Number | MFCD05022430 |
InChI Key | AGTPSAZJSOQXHJ-UHFFFAOYSA-N |
Synonym | 1-methylpiperidin-4-yl methanamine, 1-methyl-piperidin-4-methylamine, 1-methyl-4-piperidinyl methanamine, 4-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-4-yl methanamine, 1-methyl-4-aminomethyl piperidine, 4-aminomethyl-1-methylpiperidine, c-1-methyl-piperidin-4-yl-methylamine, 1-methylpiperidine-4-methylamine |
PubChem CID | 81574 |
IUPAC Name | (1-methylpiperidin-4-yl)methanamine |
SMILES | CN1CCC(CN)CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10511283
|
Thermo Scientific Maybridge
MO01153DA |
1 g | Amber Glass Bottle |
192.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10265844
|
Thermo Scientific Maybridge
MO01153DE |
5 g | Amber Glass Bottle |
755.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
7149-42-0 | |
128.22 | |
AGTPSAZJSOQXHJ-UHFFFAOYSA-N | |
81574 | |
CN1CCC(CN)CC1 |
C7H16N2 | |
MFCD05022430 | |
1-methylpiperidin-4-yl methanamine, 1-methyl-piperidin-4-methylamine, 1-methyl-4-piperidinyl methanamine, 4-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-4-yl methanamine, 1-methyl-4-aminomethyl piperidine, 4-aminomethyl-1-methylpiperidine, c-1-methyl-piperidin-4-yl-methylamine, 1-methylpiperidine-4-methylamine | |
(1-methylpiperidin-4-yl)methanamine |
Specifications
7149-42-0 | |
C7H16N2 | |
1-methylpiperidin-4-yl methanamine, 1-methyl-piperidin-4-methylamine, 1-methyl-4-piperidinyl methanamine, 4-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-4-yl methanamine, 1-methyl-4-aminomethyl piperidine, 4-aminomethyl-1-methylpiperidine, c-1-methyl-piperidin-4-yl-methylamine, 1-methylpiperidine-4-methylamine | |
CN1CCC(CN)CC1 | |
128.22 | |
128.22 | |
(1-Methyl-4-piperidinyl)methanamine |
97% | |
MFCD05022430 | |
AGTPSAZJSOQXHJ-UHFFFAOYSA-N | |
(1-methylpiperidin-4-yl)methanamine | |
81574 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title