missing translation for 'onlineSavingsMsg'
Learn More

1-Methylbenzimidazole-2-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Product Code. 11461627
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3012-80-4
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00142655
InChI Key SIRPHJCQZYVEES-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzoimidazole-2-carbaldehyde, 1-methyl-1h-benzimidazole-2-carboxaldehyde, 1-methyl-1h-benzimidazole-2-carbaldehyde, 1-methyl-1h-benzo d imidazole-2-carbaldehyde, 1-methyl-2-formylbenzimidazole, 1h-benzimidazole-2-carboxaldehyde, 1-methyl, 1-methyl-1h-1,3-benzodiazole-2-carbaldehyde, 1-methyl-1,3-benzodiazole-2-carbaldehyde, 1-methyl-2-benzimidazolecarboxaldehyde, pubchem16250
PubChem CID 762084
IUPAC Name 1-methylbenzimidazole-2-carbaldehyde
SMILES CN1C(C=O)=NC2=CC=CC=C12

Specifications

Melting Point 120°C to 122°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 160.17
Percent Purity ≥98%
Chemical Name or Material 1-Methylbenzimidazole-2-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'