missing translation for 'onlineSavingsMsg'
Learn More

1-Methylhomopiperazine, 97%, Thermo Scientific™

34.24 € - 508.00 €

Chemical Identifiers

CAS 4318-37-0
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00059810
InChI Key FXHRAKUEZPSMLJ-UHFFFAOYSA-N
Synonym n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
PubChem CID 228349
IUPAC Name 1-methyl-1,4-diazepane
SMILES CN1CCCNCC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15491678
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55441.AA
5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15401688
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55441.AC
25 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15411688
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55441.AE
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4318-37-0
114.192
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
228349
CN1CCCNCC1
C6H14N2
MFCD00059810
n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
1-methyl-1,4-diazepane

Specifications

4318-37-0
74°C to 75°C (35 mmHg)
C6H14N2
MFCD00059810
n-methylhomopiperazine, n-methyl homopiperazine, 1-methylhomopiperazine, 1h-1,4-diazepine, hexahydro-1-methyl, hexahydro-1-methyl-1h-1,4-diazepine, 1-methyl-1,4-diazaperhydroepine, methylhomopiperazine, n-methylhomopiperizine, 4-methylhomopiperazine, 1-methylhomopiperizine
CN1CCCNCC1
114.192
114.19
1-Methylhomopiperazine
0.918
45°C (113°F)
1.477
UN2920
FXHRAKUEZPSMLJ-UHFFFAOYSA-N
1-methyl-1,4-diazepane
228349
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications