missing translation for 'onlineSavingsMsg'
Learn More

1-Methylimidazole, 99%, Thermo Scientific Chemicals

44.92 € - 1115.00 €

Chemical Identifiers

CAS 616-47-7
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005292
InChI Key MCTWTZJPVLRJOU-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
PubChem CID 1390
ChEBI CHEBI:113454
IUPAC Name 1-methylimidazole
SMILES CN1C=CN=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10607482
View Documents
Thermo Scientific Acros
126991000
100 g Glass bottle
44.92 €
100g
Please to purchase this item. Need a web account? Register with us today!
10326530
View Documents
Thermo Scientific Acros
126995000
500 g Glass bottle
146.73 €
500g
Please to purchase this item. Need a web account? Register with us today!
10078531
Vedere i documenti
Thermo Scientific Acros
126990050
5 kg Plastic drum
1115.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Descrizione

Descrizione

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifica

Identificatori chimici

616-47-7
82.11
MCTWTZJPVLRJOU-UHFFFAOYSA-N
1390
1-methylimidazole
C4H6N2
MFCD00005292
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
CHEBI:113454
CN1C=CN=C1

Specifica

616-47-7
100.0
Colorless to Yellow
198.0°C
Authentic
C4H6N2
MFCD00005292
05,447
1-methyl-1h-imidazole, n-methylimidazole, 1h-imidazole, 1-methyl, imidazole, 1-methyl, n-methyl imidazole, 1-methyl-imidazole, unii-p4617qs63y, 1-methyl-1h-imidazol, n-methylimidazol, 1-methylimdazole
MCTWTZJPVLRJOU-UHFFFAOYSA-N
1-methylimidazole
1390
82.12
Liquid
98.5
-2.0°C
1.0300g/mL
92°C
98.5% min. (GC)
1.4955 to 1.4975
23, 46
1.03
Solubility in water: 145g/L (25°C). Other solubilities: soluble in organic solvents
CN1C=CN=C1
82.11
CHEBI:113454
99%
1-Methylimidazole, 99%
Videos
Sicurezza e movimentazione

Sicurezza e movimentazione

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 210-484-7

missing translation for 'rtecsNumber' : NI7000000

missing translation for 'tsca' : TSCA

Recommended Storage : this product turns yellow during storage

Identificativo prodotto
  • 1-Methylimidazole

Avvertenza
  • Danger

Categoria di pericolo
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Reproductive toxicity Category 2

Indicazioni di pericolo
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H361-Suspected of damaging fertility or the unborn child.

Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documenti

Documenti

Product Certifications

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato