missing translation for 'onlineSavingsMsg'
Learn More
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 10451084

1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 10451084
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10451084

Brand: Thermo Scientific Acros 426480010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32387-21-6
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD01321244
InChI Key HVRCLXXJIQTXHC-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole-3-carboxylic acid, 1-methyl-3-indolecarboxylic acid, 1h-indole-3-carboxylic acid, 1-methyl, 1-methylindole-3-carboxylicacid, n-methylindole-3-carboxylic acid, pubchem7285, ramosetron impurity 7, acmc-209hsw, 1-methylindole-3-carboxilic acid
PubChem CID 854040
IUPAC Name 1-methyl-1H-indole-3-carboxylic acid
SMILES CN1C=C(C(O)=O)C2=CC=CC=C12

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 197°C to 200°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 175.19
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylindole-3-carboxylic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number342636 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number426480010,426480050
compliance-icons
Product Identifier
  • 1-Methylindole-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methylindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.