missing translation for 'onlineSavingsMsg'
Learn More
1-Methylindole, 98%, Thermo Scientific Chemicals Product Code.: 11494334

1-Methylindole, 98%, Thermo Scientific Chemicals

Product Code. 11494334
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11494334

Brand: Thermo Scientific Alfa Aesar A12605.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Methylindole is useful for the determination of association constant for the electron-donor-acceptor complexes with 1-(2,4,6-trinitrophenyl) propan-2-one. It acts as a reactant for the preparation of polycyclic derivatives of indoles, bisindole derivatives and pharmaceutically active 2-oxo-1-pyrrolidine analogues. It is also used as a reactant for the preparation of Positron Emission Tomography (PET) imaging agents of protein kinase C (PKC) and glycogen synthase kinase-3 (GSK-3).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindole is useful for the determination of association constant for the electron-donor-acceptor complexes with 1-(2,4,6-trinitrophenyl) propan-2-one. It acts as a reactant for the preparation of polycyclic derivatives of indoles, bisindole derivatives and pharmaceutically active 2-oxo-1-pyrrolidine analogues. It is also used as a reactant for the preparation of Positron Emission Tomography (PET) imaging agents of protein kinase C (PKC) and glycogen synthase kinase-3 (GSK-3).

Solubility
Immiscible with water.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-76-9
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005800
InChI Key BLRHMMGNCXNXJL-UHFFFAOYSA-N
Synonym 1-methyl-1h-indole, n-methylindole, 1h-indole, 1-methyl, indole, 1-methyl, 1-methylindol, methylindole, unii-8h698roj5f, n-, chembl19912, n-methylindolShow MoreShow Less
PubChem CID 11781
IUPAC Name 1-methylindole
SMILES CN1C=CC2=CC=CC=C21

Specifications

Density 1.048
Boiling Point 237°C to 239°C
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.6065
Quantity 50 g
Beilstein 111026
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 1-Methylindole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251616 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12605.18,A12605.09
Lot Number10244796 Certificate TypeCertificate of Analysis Date28/11/2022 Catalog NumberA12605.18,A12605.09
Lot Number10143179 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12605.18,A12605.09
Lot NumberR21B060 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12605.18,A12605.09
Lot Number10155392 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12605.18,A12605.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methylindole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.