missing translation for 'onlineSavingsMsg'
Learn More

1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene, 90%, Thermo Scientific Chemicals

Product Code. 10316934
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108468-00-4
Molecular Formula C13H20N2O2
Molecular Weight (g/mol) 236.31
MDL Number MFCD02683058
InChI Key NUANLVJLUYWSER-UHFFFAOYSA-N
Synonym 1-n-boc-aminomethyl-4-aminomethyl benzene, tert-butyl 4-aminomethyl benzylcarbamate, tert-butyl n-4-aminomethyl benzyl carbamate, 4-aminomethyl-benzyl-carbamic acid tert-butyl ester, tert-butyl n-4-aminomethyl phenyl methyl carbamate, n-4-aminomethyl phenyl methyl tert-butoxy carboxamide, n-boc-p-xylylenediamine, 4-tert-butoxycarbonylaminomethyl benzylamine
PubChem CID 3354775
IUPAC Name tert-butyl N-[[4-(aminomethyl)phenyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)CN

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 64.0°C to 68.0°C
Color Beige or Pink-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 89% min. (GC)
Packaging Glass bottle
Linear Formula NH2CH2C6H4CH2NHCO2C(CH3)3
Quantity 1 g
Formula Weight 236.31
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(N-BOC-aminomethyl)-4-(aminomethyl)benzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'